Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Expedeon TruePrime™ WGA kit

Amplifies uniformly total genomic DNA from purified material.

Specifications

Product Type TruePrime™ WGA kit
Kit Type WGA kit
Polymerase Phi29 DNA polymerase and TthPrimPol
Sample Type Total genomic DNA from purified sample
Storage Requirements -20°C
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Includes Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Includes Price Quantity & Availability  
EX380100 View Documents Expedeon
380100
100 Reaction buffer (1 x 550μL), Buffer D (1 x 275μL), Buffer N (1 x 275μL), dNTPs (1 x 550μL), Water (2 x 1500μL), Enzyme 1 (1 x 550μL) and Enzyme 2 (1 x 80μL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
EX370025 View Documents Expedeon
370025
25 Reaction buffer (1 x 140μL), Buffer D (1 x 70μL), Buffer N (1 x 70μL), dNTPs (1 x 140μL), Water (1 x 750μL), Enzyme 1 (1 x 140μL) and Enzyme 2 (1 x 20μL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Uses multiple displacement amplification method
  • Consists of ten components, including the enzymes and buffers to perform the amplification reactions
  • Uses alkaline incubation to allow cell lysis and DNA denaturation of genomic DNA with very low DNA fragmentation
  • Contains Reaction buffer, Buffer D, Buffer N, dNTPs, Water, Enzyme 1 and Enzyme 2
Specifications

Specifications

TruePrime™ WGA kit
Phi29 DNA polymerase and TthPrimPol
-20°C
DNA amplification
WGA kit
Total genomic DNA from purified sample
Multiple displacement amplification method
Safety and Handling

Safety and Handling

ShelfLife : Two years

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit