Learn More
Fisherbrand™ TRUFLOW™ Tissue and Biopsy Cassettes
Just bend, snap apart, and conveniently lock
This item is not returnable.
View return policy
Description
- Hold tissue and biopsy specimens during the embedding process as well as in a storage file
- Molded from a special high-density polymer that is totally resistant to histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- One-piece integral lid eliminates the need for separate steel lids
- Can be opened or closed as often as necessary and relocked securely without danger of specimen loss
- Large labeling areas are located on three sides of the cassette
- Anterior writing area at a 30° angle
Order Info
Packaged in three dispenser boxes of 500
Specifications
Slots | |
Integral Lid | |
High Density Polymer |
30° | |
Orange | |
Tissue Cassette |