Learn More
Fisherbrand™ TRUFLOW™ Tissue and Biopsy Cassettes
Just bend, snap apart, and conveniently lock
 View more versions of this product
Catalog No. 15-200-403L
Description
Description
  • Hold tissue and biopsy specimens during the embedding process as well as in a storage file
  • Molded from a special high-density polymer that is totally resistant to histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Can be opened or closed as often as necessary and relocked securely without danger of specimen loss
  • Large labeling areas are located on three sides of the cassette
  • Anterior writing area at a 30° angle
Order Info
Packaged in three dispenser boxes of 500
Specifications
Specifications
Slots
Integral Lid
High Density Polymer
30°
Peach
Tissue Cassette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.