Learn More
Aat BioquestSupplier Diversity Partner Trypan Blue, sodium salt *UltraPure grade* *Purified to eliminate fluorescent impurities*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2452
Trypan blue, a diazo dye is a vital stain that colors dead tissues or cells blue. Live cells or tissues with intact cell membranes are not be colored.
Catalog No. 50-195-3170