Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trypsin 1/PRSS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244111
Description
Specifically detects Trypsinogen in Bovine samples, and it is validated for ELISA, Western Blotting
Trypsin 1/PRSS1 Polyclonal specifically detects Trypsin 1/PRSS1 in Bovine samples. It is validated for Western Blot, ELISA.Specifications
Trypsin 1/PRSS1 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A | |
Rabbit | |
25.79 kDa | |
100 μg | |
Cancer | |
5644 | |
Reconstitute with 100 ul deionized water (or equivalent) | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 1:1000-1:5000, ELISA 1:5000-1:20000 | |
P00760 | |
PRSS1 | |
Trypsin 1/PRSS1 Antibody was produced by repeated immunizations with bovine pancreatic trypsin protein purified. (Uniprot: P00760) | |
Multi-step | |
RUO | |
Primary | |
Cross-reactivity against Trypsin 1/PRSS1 from other tissues and species may occur but has not been specifically determined. | |
Bovine, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction