Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin 1/PRSS1 Mouse anti-Human, Bovine, Alexa Fluor™ 532, Clone: 7C8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223517AF532

Catalog No. NP22317A32


Add to cart

Description

Description

Trypsin 1/PRSS1 Monoclonal antibody specifically detects Trypsin 1/PRSS1 in Human, Bovine samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Trypsin 1/PRSS1
7C8
Western Blot, ELISA
Conjugated, Purified
PRSS1
Trypsin purified from human pancreatic tissue and adsorbed onto aluminum hydroxide gel (2)
Protein A or G purified
Cancer
Primary
5644.0
Bovine, Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Clone 7C8 is specific for human trypsin, trypsinogen and trypsin in complex with alpha1-antitrypsin. Clone 7C8 cross-reacts weakly with bovine trypsin.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit