Learn More
Trypsin 1/PRSS1 Mouse anti-Human, Bovine, Alexa Fluor™ 750, Clone: 7C8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP223517AF750
Catalog No. NP22317A70
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Trypsin 1/PRSS1 Monoclonal antibody specifically detects Trypsin 1/PRSS1 in Human,Bovine samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Trypsin 1/PRSS1
7C8
Western Blot, ELISA
Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 7C8 is specific for human trypsin, trypsinogen and trypsin in complex with alpha1-antitrypsin. Clone 7C8 cross-reacts weakly with bovine trypsin.
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
PRSS1
Trypsin purified from human pancreatic tissue and adsorbed onto aluminum hydroxide gel (2)
0.1 mL
Primary
5644
Bovine, Human