Learn More
BioworldSupplier Diversity Partner Trypsin-Digestion Mix, 100 Prep(s) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220400591
Trypsin Digestion Buffer Used for efficient digestion of proteins with trypsin. Modified Mass- Spec grade for use with the In-gel extraction kit. Sufficient for 100 in gel digestions.
Catalog No. 50-253-067