Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Trypsin Inhibitor, soybeans

Catalog No. p-7082253
Click to view available options
Quantity:
100 mg
1 g

Inhibits the pancreatic enzyme trypsin | CAS: 9035-81-8

The soybean trypsin inhibitor has been tested for cell culture applications. It has been used in a study to assess the potential application in animal cell culture of an alkaline protease from a non-toxigenic mangrove isolate of Vibri sp. V26. It has also been used in a study to improve the detection of fungi in eosinophilic mucin. It is an inhibitor of trypsin factor Xa, plasmin and neutrophil elastase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Dissolve in 1X PBS at 1 to 10mg/ml for stock solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9035-81-8

Specifications

Quantity 1 g
Description Off-white to Pale Brown
Content And Storage 2°C to 8°C
For Use With (Application) Inhibits the pancreatic enzyme trypsin
Product Type Signal Transduction Reagent
Form Powder
EINECSNumber 232-906-9
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.