Learn More
Trypsin Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Trypsin |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0809
![]() ![]() |
Rockland Immunochemicals
2004011390100 |
100 μg |
Each for $636.24
|
|
|||||
50-199-0810
![]() ![]() |
Rockland Immunochemicals
200401139S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Trypsin Antibody is an IgG fraction purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Trypsin (Bovine Pancreas). Cross-reactivity against Trypsin from other tissues and species may occur but has not been specifically determined.
Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.Specifications
Trypsin | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Bovine | |
Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsin I; Anionic trypsin II; Anionic trypsin-1; Anionic trypsin-2; Anionic trypsinogen; Beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; pancreatic trypsin 2; pancreatic trypsin II; Pretrypsinogen I; Pretrypsinogen II; Protease Serine 1; protease serine 2 preproprotein; protease, serine 1; protease, serine 1 (trypsin 1); protease, serine 2; protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; protease, serine, 2 (trypsin 2); Prss1; PRSS2; PTRYI; Ptryss2; RATPTRYI; Serine protease 1; Serine protease 2; Ta; TCR V beta 4.1; Tesp4; TRP1; TRY1; Try-1; TRY2; TRY4; TRY8; Trygn16; TRYP; TRYP1; TRYP2; TRYP8; trypsin 1; trypsin 2; Trypsin I; Trypsin II; Trypsin-1; Trypsin-2; trypsinogen 1; trypsinogen 16; trypsinogen 2; trypsinogen 20; trypsinogen A | |
PRSS1, PRSS2 | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
282603, 615026 | |
Anti-Trypsin Antibody was produced by repeated immunizations with bovine pancreatic trypsin protein purified. | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
PRSS2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.