Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin, porcine pancreas

A pancreatic serine protease

$71.20 - $252.00

Specifications

CAS 9002-07-7
Molecular Weight 23.8 kd
Synonym EC 3.4.21.4; Trypsin IV porcine
Storage Requirements −30° to −10°C
Format Lyophilized Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6040209 View Documents Alfa Aesar™
J6040209
10g Each for $71.20
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ6040218 View Documents Alfa Aesar™
J6040218
50g Each for $252.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

Reconstitute in aqueous buffer
Specifications

Specifications

9002-07-7
EC 3.4.21.4; Trypsin IV porcine
Lyophilized Powder
Off-white to Pale Brown
23.8 kd
−30° to −10°C
A pancreatic serine protease
Trypsin
Safety and Handling

Safety and Handling

EINECSNumber : 232-650-8

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit