Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptamine, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
61-54-1
C10H12N2
160.22
APJYDQYYACXCRM-UHFFFAOYSA-N
tryptamine,2-1h-indol-3-yl ethanamine,3-2-aminoethyl indole,1h-indole-3-ethanamine,2-3-indolyl ethylamine,indol-3-ethylamine,indole, 3-2-aminoethyl,2-indol-3-yl ethylamine,2-1h-indol-3-yl ethan-1-amine,tryptamin
1150
CHEBI:16765
2-(1H-indol-3-yl)ethanamine
C1=CC=C2C(=C1)C(=CN2)CCN
61-54-1
C10H12N2
160.22
APJYDQYYACXCRM-UHFFFAOYSA-N
tryptamine,2-1h-indol-3-yl ethanamine,3-2-aminoethyl indole,1h-indole-3-ethanamine,2-3-indolyl ethylamine,indol-3-ethylamine,indole, 3-2-aminoethyl,2-indol-3-yl ethylamine,2-1h-indol-3-yl ethan-1-amine,tryptamin
1150
CHEBI:16765
2-(1H-indol-3-yl)ethanamine
C1=CC=C2C(=C1)C(=CN2)CCN
Water 2% Max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Boiling Point 137.0°C (0.1 mmHg)
Flash Point 185°C
Melting Point 113.0°C to 117.0°C
Appearance (1% soln. in MeOH) clear, yellowish
Quantity 250g
Assay Percent Range 97.5% min. (GC)
Beilstein 22,I,636
Merck Index 15,9976
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ethanol and acetone, practically insoluble in ether, benzene and, chloroform
Formula Weight 160.22
Physical Form Flakes or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Tryptamine
Show More Show Less

EINECSNumber : 200-510-5