Alfa Aesar™ Tryptamine hydrochloride, 98+%

Manufacturer: Alfa Aesar™ A1103030

Quantity 250g

 View more versions of this product

Catalog No. AAA1103030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 343-94-2
    Molecular Formula C10H12N2·HCl
    Formula Weight 196.68
    MDL Number MFCD00012682
    Beilstein 3568419
    Merck Index 14,9796
    Synonym 3-(2-Aminoethyl)indole hydrochloride; 2-(3-Indolyl)ethylamine hydrochloride
    Chemical Name or Material Tryptamine hydrochloride

    Specifications

    Formula Weight 196.68
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Molecular Formula C10H12N2·HCl
    Merck Index 14,9796
    Synonym 3-(2-Aminoethyl)indole hydrochloride; 2-(3-Indolyl)ethylamine hydrochloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    RTECS Number NL4375000
    TSCA TSCA
    MDL Number MFCD00012682
    CAS 343-94-2
    Beilstein 3568419
    Melting Point 252° to 255°C
    Quantity 250g
    Chemical Name or Material Tryptamine hydrochloride
    Assay ≥98%

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit