Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/TPS1, Mouse anti-Human, Alexa Fluor 647, Clone: TPSAB1/1961, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275754AF647

Catalog No. NB008880


Add to cart

Description

Description

Tryptase alpha/TPS1 Monoclonal antibody specifically detects Tryptase alpha/TPS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Tryptase alpha/TPS1
TPSAB1/1961
Immunohistochemistry, Immunohistochemistry-Paraffin
50mM Sodium Borate with 0.05% Sodium Azide
TPSAB1
Recombinant human Tryptase protein fragment (around aa 115-233) (Exact sequence is proprietary)
Monoclonal
Protein A or G purified
RUO
Human
Immunohistochemistry,Immunohistochemistry (Paraffin)
Alexa Fluor 647
Purified,Conjugated
TPS1, TPS2, TPSAB1, TPSB1, tryptase alpha/beta 1
Mouse
IgG1 Kappa
Primary
0.1 mL
Store at 4°C in the dark.
Tryptases comprise a family of trypsin-like serine proteases (peptidase family S1). Tryptases are stored in mast cell secretory granules and basophils. Mast cells are connective tissue cells derived from blood-forming tissues that line arterial walls and secrete substances, which mediate inflammatory and immune responses. Tryptases are released into the extracellular environment and are resistant to all known endogenous proteinase inhibitors. This antibody reacts with mast cells distributed in skin, synovium, lung, and heart. This antibody does not bind with any other cell type. Human mast cell tryptase is considered to be an important marker of mast cell activation and is an important mediator of inflammation. Mastocytosis is a term collectively used for a group of disorders in which there is abnormal accumulation of mast cells in one or multiple organs. Anti-tryptase, combined with anti-CD2, anti-CD25, and anti-CD117, can be useful in identifying reactive mast cell hyperplasia, myelogenous neoplasms, mast cell leukemia, and mastocytosis.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit