Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/TPS1 Mouse anti-Human, Clone: TPSAB1/1963, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Tryptase alpha/TPS1
Clone TPSAB1/1963
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122870
View Documents
Novus Biologicals
NBP30769820UG
20 μg
Each for $138.37
Add to cart
 
NB122869
View Documents
Novus Biologicals
NBP30769810UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Tryptase alpha/TPS1 Monoclonal antibody specifically detects Tryptase alpha/TPS1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Tryptase alpha/TPS1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
7177
IgG1 Kappa
Primary
Store at 4°C.
TPSAB1/1963
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human
TPS1, TPS2, TPSAB1, TPSB1, tryptase alpha/beta 1
Recombinant human Tryptase alpha/TPS1 protein fragment (around aa 115-233) (Exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit