Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase alpha/TPS1, Mouse anti-Human, DyLight 350, Clone: TPSAB1/1961, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275754UV
Description
Tryptase alpha/TPS1 Monoclonal antibody specifically detects Tryptase alpha/TPS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Tryptase alpha/TPS1 | |
TPSAB1/1961 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TPSAB1 | |
Recombinant human Tryptase protein fragment (around aa 115-233) (Exact sequence is proprietary) | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Human |
Immunohistochemistry,Immunohistochemistry (Paraffin) | |
DyLight 350 | |
Purified,Conjugated | |
TPS1, TPS2, TPSAB1, TPSB1, tryptase alpha/beta 1 | |
Mouse | |
IgG1 Kappa | |
Primary | |
0.1 mL | |
Store at 4°C in the dark. | |
Tryptases comprise a family of trypsin-like serine proteases (peptidase family S1). Tryptases are stored in mast cell secretory granules and basophils. Mast cells are connective tissue cells derived from blood-forming tissues that line arterial walls and secrete substances, which mediate inflammatory and immune responses. Tryptases are released into the extracellular environment and are resistant to all known endogenous proteinase inhibitors. This antibody reacts with mast cells distributed in skin, synovium, lung, and heart. This antibody does not bind with any other cell type. Human mast cell tryptase is considered to be an important marker of mast cell activation and is an important mediator of inflammation. Mastocytosis is a term collectively used for a group of disorders in which there is abnormal accumulation of mast cells in one or multiple organs. Anti-tryptase, combined with anti-CD2, anti-CD25, and anti-CD117, can be useful in identifying reactive mast cell hyperplasia, myelogenous neoplasms, mast cell leukemia, and mastocytosis. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title