Learn More
Thermo Scientific™ Tryptic Soy Broth Available on GSA/VA Contract for Federal Government customers only.
Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.
Manufacturer:  Thermo Scientific™ R065002
 View more versions of this product
Catalog No. R065002
Add to cart
This item is not returnable. View return policy
Description
Description
Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use TSB tubes.
Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
20/Pk.
United States
Broth
15mm x 103mm Tube
4.5 mL
Tryptic Soy Broth