Learn More
Research Products International CorpSupplier Diversity Partner Tryptone, Granulated, 5 Kilograms DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner T600655000.0
Pancreatic digest of casein. Material is specially granulated to reduce dust and increase solubility rate. Ingredients: Total Nitrogen (TN): ~12.7% Amino Nitrogen (AN): ~3.7% AN/TN Ratio: 0.26 Sodium Chloride: ~0.4% Tryptophan: 1.0 pH (1% solution): ~7.3%
Catalog No. 50-489-114