Learn More
BioVision IncSupplier Diversity Partner Tryptophan Assay Kit (Fluorometric): Tryptophan Assay Kit (Fluorometric): Convenient assay to measure tryptophan in serum, urine or other biological fluids. Detection limit: 2.5 µM. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K557100
Tryptophan Assay Kit (Fluorometric): Tryptophan (TRP) is one of the eight essential amino acids. Changes in tryptophan concentrations are directly related to a number of physiological and behavioral processes including: sleep, memory, depression, motion sickness, bipolar disorders, and schizophrenia. BioVision's Tryptophan Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects tryptophan concentration in biological fluids, including free and bound tryptophan in serum. The principle is based on the non-enzymatic reaction to generate a fluorophore (Ex/Em = 370/440 nm) using tryptophan as a building block. The reaction is specific and other amino acids do not interfere with the assay. The assay can detect as little as 2.5 ¿M of TRP in a variety of biological samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9770