Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptophol 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
526-55-6
C10H11NO
161.2
MFCD00005659
MBBOMCVGYCRMEA-UHFFFAOYSA-N
tryptophol,3-2-hydroxyethyl indole,indole-3-ethanol,1h-indole-3-ethanol,2-1h-indol-3-yl ethanol,3-indoleethanol,indoleethanol,3-indolylethanol,indole ethanol,2-3-indolyl ethanol
10685
CHEBI:17890
2-(1H-indol-3-yl)ethanol
C1=CC=C2C(=C1)C(=CN2)CCO
526-55-6
C10H11NO
161.2
MFCD00005659
MBBOMCVGYCRMEA-UHFFFAOYSA-N
tryptophol,3-2-hydroxyethyl indole,indole-3-ethanol,1h-indole-3-ethanol,2-1h-indol-3-yl ethanol,3-indoleethanol,indoleethanol,3-indolylethanol,indole ethanol,2-3-indolyl ethanol
10685
CHEBI:17890
2-(1H-indol-3-yl)ethanol
C1=CC=C2C(=C1)C(=CN2)CCO
Quantity 5g
Color White to Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 57.0°C to 60.0°C
Boiling Point 174.0°C (2.7 mbar)
Flash Point 255°C
Assay Percent Range 97%
Beilstein 21, I, 218
Merck Index 15, 9978
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, ethanol, acetone, chloroform, and ethyl acetate
Formula Weight 161.2
Physical Form Crystalline Mass, Crystals, Flakes or Powder
Percent Purity 97%
Chemical Name or Material Tryptophol, 97%
Show More Show Less

EINECSNumber : 208-393-2

RTECSNumber : KL3685000

TSCA : TSCA