Learn More
BioworldSupplier Diversity Partner Tryptose, 500 g DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306200891
TryptoseSynonym: TrypticaseEnzymatic digest of protein for use in preparing microbiological culture mediapH (1% Solution): 7.0Dehydrated Appearance: light beige, homogeneous powderPlant tissue culture testedSoluble in water
Catalog No. 50-254-388