Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSEN2 Polyclonal Antibody
TSEN2 Polyclonal Antibody
Supplier: Thermo Scientific PA555513
Description
TSEN2 Polyclonal Antibody for Western Blot, IHC

Specifications
TSEN2 | |
Polyclonal | |
Unconjugated | |
Tsen2 | |
AU067695; HsSen2; PCH2B; RGD1309946; SEN2; SEN2L; tRNA splicing endonuclease 2 homolog (S. cerevisiae); tRNA splicing endonuclease subunit 2; tRNA-intron endonuclease Sen2; tRNA-intron nuclease 2; tRNA-splicing endonuclease subunit Sen2; Tsen2; TSEN2 tRNA splicing endonuclease subunit | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCE0 | |
Tsen2 | |
Recombinant protein corresponding to Human TSEN2. Recombinant protein control fragment (Product #RP-94795). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction