Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSEN2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549147

Catalog No. PA549147

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Constitutes one of the two catalytic subunit of the tRNA-splicing endonuclease complex, a complex responsible for identification and cleavage of the splice sites in pre-tRNA. It cleaves pre-tRNA at the 5 ft.- and 3 ft.-splice sites to release the intron. The products are an intron and two tRNA half-molecules bearing 2 ft.,3 ft.-cyclic phosphate and 5 ft.-OH termini. There are no conserved sequences at the splice sites, but the intron is invariably located at the same site in the gene, placing the splice sites an invariant distance from the constant structural features of the tRNA body. Isoform 1 probably carries the active site for 5 ft.-splice site cleavage. The tRNA splicing endonuclease is also involved in mRNA processing via its association with pre- mRNA 3 ft.-end processing factors, establishing a link between pre- tRNA splicing and pre-mRNA 3 ft.-end formation, suggesting that the endonuclease subunits function in multiple RNA-processing events. Isoform 2 is responsible for processing a yet unknown RNA substrate. The complex containing isoform 2 is not able to cleave pre-tRNAs properly, although it retains endonucleolytic activity.
Specifications

Specifications

TSEN2
Polyclonal
PBS with 0.09% sodium azide
Q8NCE0
tRNA-splicing endonuclease subunit Sen2, tRNA-intron endonuclease Sen2, HsSen2, TSEN2, SEN2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80746
Western Blot
Unconjugated
TSEN2
Liquid
TSEN2
KLH conjugated synthetic peptide between 16-43 amino acids from the N-terminal region of human TSEN2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit