Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSG Rat anti-Mouse, DyLight 488, Clone: RM0140-7D19, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122516G
Description
TSG Monoclonal antibody specifically detects TSG in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TSG | |
RM0140-7D19 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Purified | |
TWSG1 | |
Mouse recombinant protein of TSG (full length). | |
Protein G purified | |
Store at 4C in the dark. | |
Monoclonal | |
Mouse |
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TSGtwisted gastrulation protein homolog 1, twisted gastrulation homolog 1 (Drosophila) | |
Rat | |
IgG2 | |
0.1 ml | |
Primary | |
57045 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title