Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ TSG Series General Purpose Laboratory Refrigerators
Designed for your everyday laboratory needs
Supplier: Thermo Scientific™ TSG49RPSA
Description
Thermo Scientific™ TSG Series general-purpose laboratory refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2° to 10°C controlled environment.
- Featuring a wide range of cabinet sizes from 12 cu. ft. to 72 cu. ft.
- Whisper-quiet operation
- ENERGY STAR certifiedTemperature, door ajar*, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
- Warranty: varies by country, please contact us for details
*All models except TSG45RPLA and TSG45RSLA come equipped with door ajar alarm

Specifications
TSG Series general purpose laboratory refrigerator | |
1387 L | |
+5°C | |
Solid | |
Stainless Steel | |
100 lb. | |
White-painted | |
Stainless Steel | |
Digital Microprocessor | |
2 years parts and labor, plus 3-year compressor warranty | |
Alarm | |
28.5x49x61.75 in. (724x1245x1569 mm) | |
8.8 A | |
115 V | |
60 Hz | |
597 lb. |
49 cu. ft. | |
2°C to 10°C | |
R290 | |
2 | |
8 Full | |
45 kg | |
Painted | |
cULus, ENERGYSTAR, Prop 65 | |
1 Rear access port | |
Adaptive Defrost | |
Key Lock | |
32x52.7x81 in.(813x1339x2057 mm) | |
NEMA 5-15 P | |
115 V 60 Hz | |
Energy efficient | |
271 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction