Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ TSG Series General Purpose Laboratory Refrigerators
Designed for your everyday laboratory needs
Supplier: Thermo Scientific™ TSG25RSSA
Description
Thermo Scientific™ TSG Series general-purpose laboratory refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2° to 10°C controlled environment.
- Featuring a wide range of cabinet sizes from 12 cu. ft. to 72 cu. ft.
- Whisper-quiet operation
- ENERGY STAR certifiedTemperature, door ajar*, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
- Warranty: varies by country, please contact us for details
*All models except TSG45RPLA and TSG45RSLA come equipped with door ajar alarm
Specifications
TSG Series general purpose laboratory refrigerator | |
651 L | |
+5°C | |
Solid | |
Stainless Steel | |
100 lb. | |
Gray-painted | |
Stainless Steel | |
Digital Microprocessor | |
2 years parts and labor, plus 3-year compressor warranty | |
Alarm | |
28.5x24x61.75 in. (724x610x1569 mm) | |
5.8 A | |
115 V | |
60 Hz | |
392 lb. |
23 cu. ft. | |
2°C to 10°C | |
R290 | |
1 | |
4 Full | |
45 kg | |
Painted | |
cULus, ENERGYSTAR, Prop 65 | |
1 Rear access port | |
Adaptive Defrost | |
Key Lock | |
32x27.75x81 in.(813x705x2057 mm) | |
NEMA 5-15 P | |
115 V 60 Hz | |
Energy efficient | |
175 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction