Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSH beta Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TSH112AP
Description
TSH beta Polyclonal antibody specifically detects TSH beta in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western BlotSpecifications
TSH beta | |
Polyclonal | |
Unconjugated | |
TSHB | |
thyroid stimulating hormone beta; thyroid stimulating hormone subunit beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone beta subunit; thyroid-stimulating hormone subunit beta; Thyrotropin alfa; Thyrotropin beta chain; thyrotropin beta subunit; thyrotropin subunit beta; TSH; Tshb; TSH-B; TSH-beta | |
Synthetic peptide corresponding to unique amino acid sequence on human TSH beta chain protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P01222, P04652, P12656 | |
Rabbit | |
Affinity chromatography | |
RUO | |
22094, 25653, 7252 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction