Learn More
Drg International IncSupplier Diversity Partner Tsh Elisa DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA1782
An enzyme immunoassay for the quantitative determination of thyroid stimulating hormone (TSH) in serum. The determination of serum or plasma levels of thyroid stimulating hormone (TSH or thyrotropin) is recognized as a sensitive method in the diagnosis of primary and secondary hypothyroidism. The TSH ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay. The assay system utilizes a unique monoclonal antibody directed against a distinct antigenic determinant on the intact TSH molecule. Mouse monoclonal anti-TSH antibody is used for solid phase immobilization (microtiter wells), and goat anti-TSH antibody is in the antibody-enzyme (horseradish peroxidase) conjugate solution.
Catalog No. 50-194-7539