Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHR Mouse anti-Human, Allophycocyanin/Cy7, Clone: 4C1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB10065797APCCY7

Catalog No. N1065797AP7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TSHR Monoclonal antibody specifically detects TSHR in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TSHR
4C1
Flow Cytometry
Conjugated, Purified
TSHR
External domain of human TSH receptor.
Protein A or G purified
Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience
Primary
7253.0
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Recognizes the human thyroid stimulating hormone (TSH) receptor (otherwise known as the thyrotropin receptor) binding to an extracellular domain.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit