Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSHR Antibody (TSHRB/1404) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254395100UG
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
TSH R | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready | |
CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
Mouse | |
42 kDa | |
100 μg | |
Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience | |
7253 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
TSHRB/1404 | |
Unconjugated | |
PBS with No Preservative | |
TSHR | |
Recombinant human TSHR, B-Chain protein | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction