Learn More
TSHZ1 Mouse anti-Human, Clone: OTI8B9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809980
Catalog No. 50-166-9827
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Teashirt zinc finger homeobox (TSHZ) family comprise a family of evolutionarily conserved transcription factors that, in Drosophila, are active in specific body parts for patterning, but whose function in vertebrates is less clear. TSHZ1 has been found to be required for axial skeleton, soft palate and middle ear development in mice and may be involved in a common pathway with the Hox genes. Both TSHZ1 and the related protein TSHZ3 have been found to interact with FE65, an adapter protein that binds to the amyloid protein precursor (APP) in neurons. Together with SET, a component of the inhibitor of acetyl transferase, and histone deacetylases, these proteins formed a gene-silencing complex whose target includes caspase-4.
Specifications
Specifications
TSHZ1
Monoclonal
Unconjugated
TSHZ1
Lyophilized
TSHZ1
Human recombit protein fragment corresponding to amino acids 29-176 of human TSHZ1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8B9
PBS with 8% trehalose and no preservative; pH 7.3
Q6ZSZ6
CAA, NY-CO-33, SDCCAG33, TSH1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10194