Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSLP Mouse anti-Human, Alexa Fluor™ 350, Clone: 55N1D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227326AF350

Catalog No. NP227326A30


Add to cart

Description

Description

TSLP Monoclonal antibody specifically detects TSLP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TSLP
55N1D10
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TSLP
A partial recombinant TSLP protein (amino acids 27-158) was used as the immunogen for the antibody.
Protein G purified
Biologically Active Proteins, Innate Immunity
Primary
85480.0
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
thymic stromal lymphopoietin
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit