Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSLP Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems AF1398SP
This item is not returnable.
View return policy
Description
TSLP Polyclonal specifically detects TSLP in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
TSLP | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
thymic stromal lymphopoietin | |
Sheep | |
Affinity Purified | |
RUO | |
85480 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.05-0.25 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q969D9 | |
TSLP | |
E. coli-derived recombinant human TSLP Tyr29-Gln159 Accession # Q969D9 | |
25 μg | |
Primary | |
Detects human TSLP in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant mouse TSLP and recombinant human TSLP R is observed. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction