Learn More
TSLPR/CRLF2 Mouse anti-Human, Alexa Fluor™ 532, Clone: 59N5G4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227331AF532
Catalog No. NP227331A32
Add to cart
Description
Description
TSLPR/CRLF2 Monoclonal antibody specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
TSLPR/CRLF2
59N5G4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
TSLPR is a glycoprotein with two major isoforms of 359 amino acids and 255 amino acids. In western blotting, bands may be visualized between 27 kD and ~50 kD.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CRLF2
Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
0.1 mL
Primary
64109
Human