Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSP23 Mouse anti-Human, Mouse, Clone: Tsp232A, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527583

Catalog No. PIMA527583


Add to cart

Description

Description

p23 is a highly conserved ubiquitous protein, known to have an important function as a cochaperone for the HSP90 chaperoning system. Studies have revealed that p23 is a small protein (18 to 25 kDa) with a simple structure. p23 is a phosphor-protein, which is highly acidic and has an aspartic acid-rich c-terminal domain. Numerous studies have found p23 to be associated with other client proteins like Fes tyrosine kinase, the heme regulated kinase HRI, hsf1 transcription factor, aryl hydrocarbon receptor, telomerase, and Hepadnavirus reverse transcriptase. In spite of several years of study, the exact functional significance of p23 is still not clear. p23 is thought to be involved in the adenosine triphosphate-mediated HSP90 binding of client proteins. Since many HSP90 client proteins are involved in oncogenic survival signaling, a recent study has concluded p23 to be a promising target in leukemic apoptosis. HSP90 and its co-chaperone p23 are certainly among the emerging anti-tumor targets in oncology. Specifically TSP23 (transcript similar p23) displays 44% and 17% amino acid identity with p23 and Sba1p respectively.
Specifications

Specifications

TSP23
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Q3THG9, Q9BTE6
Transcript similar p23, AARSD1, AASD1_HUMAN, Alanyl-tRNA editing protein Aarsd1, Alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744, OTTHUMP00000204017, OTTHUMP00000204019, OTTHUMP00000204020
Mouse
IgG1, kappa
100 μg
-20°C
Primary
69684, 80755
Immunohistochemistry (PFA fixed), Western Blot
Tsp232A
Unconjugated
AARSD1
Liquid
Aarsd1
Recombit tsp23
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit