Learn More
Aviva Systems Biology Corp TSPAN10 Antibody - N-terminal region (ARP50022_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50022_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TSPAN10. Alternate names: FLJ39607, OCSPApplicationWB. Species reactivity: Human
Catalog No. 50-014-60243