Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542947
Description
Peptide sequence: EFPGCSKQAH QEDLSDLYQE GCGKKMYSFL RGTKQLQVLR FLGISIGVTQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
TSPAN12 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications
TSPAN12 | |
Polyclonal | |
Unconjugated | |
TSPAN12 | |
9030619E17; AI426782; AI663988; AW111457; EVR5; MGC137611 protein; NET2; NET-2; Tetraspan NET-2; tetraspanin 12; tetraspanin-12; Tm4sf12; transmembrane 4 superfamily member 12; TSPAN12; Tspan-12; UNQ774/PRO1568; zgc:63880 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23554 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O95859 | |
TSPAN12 | |
Synthetic peptide directed towards the middle region of human TSPAN12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction