Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550891

Catalog No. PA550891


Add to cart

Description

Description

The antibody detects endogenous levels of total TSPAN13 / TM4SF13 protein.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications

Specifications

TSPAN13
Polyclonal
Unconjugated
TSPAN13
Liquid
TSPAN13
Synthetic peptide corresponding to a region derived from internal residues of human tetraspanin 13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
2.1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O95857, Q9D8C2, Q5FVL6
NET6, TM4SF13
Rabbit
IgG
100 μL
-20°C
Primary
27075, 366602, 66109
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit