Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN18 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548957

Catalog No. PA548957


Add to cart

Specifications

Specifications

TSPAN18
Polyclonal
Unconjugated
TSPAN18
Liquid
TSPAN18
KLH conjugated synthetic peptide between 93-120 amino acids from the Central region of human TSPAN18.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q96SJ8
Tetraspanin-18, Tspan-18, TSPAN18
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90139
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit