Learn More
TSPAN2 Mouse anti-Human, Alexa Fluor 750, Clone: 822509, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB7876S100UG
Catalog No. FAB7876S100
Add to cart
This item is not returnable. View return policy
Description
Description
TSPAN2 Monoclonal antibody specifically detects TSPAN2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TSPAN2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
O60636
TSPAN2
NS0 mouse myeloma cell line transfected with human TSPAN2 Met1-Ile221 Accession # O60636
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human TSPAN2 in ELISAs.
Flow Cytometry
822509
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FLJ12082, NET3, new EST tetraspan 3, tetraspan 2, Tetraspan NET-3, tetraspan TM4SF, tetraspanin 2, tetraspanin 2 isoform, tetraspanin-2, TSN2, Tspan-2
Mouse
IgG2b
RUO
Primary
10100
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)