Learn More
TSPAN32 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 203201AP
Catalog No. 50-173-2826
Add to cart
Description
Description
This gene, which is a member of the tetraspanin superfamily, is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of chromosome 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian and breast cancers. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4, escapes imprinting. This gene may play a role in maligcies and diseases that involve this region, and it is also involved in hematopoietic cell function. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
Specifications
TSPAN32
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96QS1, Q9JHH2
PHEMX, PHMX, Protein Phemx, tetraspanin 32, Tspan 32, TSPAN32, TSSC6
Rabbit
IgG
150 μL
-20°C
Primary
10077, 27027
Western Blot
Unconjugated
TSPAN32
Liquid
TSPAN32
TSPAN32 Fusion Protein Ag14174
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse