Learn More
TSPAN7/TM4SF2 Mouse anti-Human, Alexa Fluor 350, Clone: 482618, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB9179U100UG
Catalog No. FAB9179U100
Add to cart
This item is not returnable. View return policy
Description
Description
TSPAN7/TM4SF2 Monoclonal antibody specifically detects TSPAN7/TM4SF2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TSPAN7/TM4SF2
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
A15, A15CCG-B7, CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, DXS1692E, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, TALLA-1, T-cell acute lymphoblastic leukemia-associated antigen 1, tetraspanin 7, tetraspanin-7, TM4SF2, TM4SF2b, TM4SF2tetraspanin protein, transmembrane 4 superfamily 2b, Transmembrane 4 superfamily member 2X chromosome, surface marker 1, transmembrane protein A15, Tspan-7
HEK293 human embryonic kidney cell line transfected with human TSPAN7 Accession # P41732
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains human TSPAN7 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
482618
Flow Cytometry 0.25-1 ug/10^6 cells
P41732
Mouse
IgG2a
RUO
Primary
7102
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)