Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN8 Rat anti-Mouse, BB700, Clone: 657909, BD OptiBuild
Rat Monoclonal Antibody
Manufacturer: BD Biosciences 748044
Description
TSPAN8 Monoclonal antibody specifically detects TSPAN8 in Mouse samples. It is validated for Flow Cytometry.Specifications
TSPAN8 | |
657909 | |
BB700 | |
Rat | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal | |
Mouse |
Flow Cytometry | |
0.2 mg/mL | |
Conjugated, Purified | |
IgG2b, κ | |
RUO | |
Primary | |
TSPAN8 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title