Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN8/TM4SF3 Rat anti-Human, Alexa Fluor 647, Clone: 458811, R&D Systems™

Rat Monoclonal Antibody

Manufacturer:  R&D Systems FAB4734R100UG

Catalog No. FAB4734R100


Add to cart
This item is not returnable. View return policy

Description

Description

TSPAN8/TM4SF3 Monoclonal specifically detects TSPAN8/TM4SF3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TSPAN8/TM4SF3
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
AAH05246
TSPAN8
NS0 mouse myeloma cell line transfected with human TSPAN8 Met1-Lys237 (Ile35Val) Accession # AAH05246
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7103
Reconstitution default not known
Flow Cytometry
458811
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CO-029, tetraspanin 8, TM4SF3, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029
Rat
IgG2b
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human TSPAN8 in Western blots.
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)