Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPEAR Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550349

Catalog No. PA550349


Add to cart

Description

Description

The antibody detects endogenous levels of total TSPEAR protein.

This gene encodes a protein that contains a N-terminal thrombospondin-type laminin G domain and several tandem arranged epilepsy-associated repeats (EARs). A mutation in this gene is the cause of autosomal recessive deafness-98. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

TSPEAR
Polyclonal
Unconjugated
TSPEAR
Liquid
TSPEAR
Fusion protein corresponding to a region derived from internal residues of human thrombospondin-type laminin G domain and EAR repeats
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.4 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8WU66
C21orf29
Rabbit
IgG
100 μL
-20°C
Primary
54084
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit