Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPY1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548721

Catalog No. PA548721


Add to cart

Description

Description

May be involved in sperm differentiation and proliferation.
Specifications

Specifications

TSPY1
Polyclonal
PBS with 0.09% sodium azide
Q01534
Testis-specific Y-encoded protein 1, Cancer/testis antigen 78, CT78, TSPY1, TSPY
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7258
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TSPY1
Liquid
TSPY1
KLH conjugated synthetic peptide between 274-303 amino acids from the C-terminal region of human TSPY.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit