Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPYL5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548599

Catalog No. PA548599


Add to cart

Description

Description

Involved in modulation of cell growth and cellular response to gamma radiation probably via regulation of the Akt signaling pathway. Involved in regulation of p53/TP53. Suppresses p53/TP53 protein levels and promotes its ubiquitination; the function is dependent on USP7 and independent on MDM2. Proposed to displace p53/TP53 from interaction with USP7.
Specifications

Specifications

TSPYL5
Polyclonal
PBS with 0.09% sodium azide
Q86VY4, Q69ZB3
Testis-specific Y-encoded-like protein 5, TSPY-like protein 5, TSPYL5, KIAA1750
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
239364, 85453
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TSPYL5
Liquid
TSPYL5
KLH conjugated synthetic peptide between 344-372 amino acids from the C-terminal region of human TSPYL5.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit