Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPYL6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15286920UL

 View more versions of this product

Catalog No. NBP15286920


Add to cart

Description

Description

TSPYL6 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

TSPYL6
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
TSPYL6
Synthetic peptides corresponding to TSPYL6 (TSPY-like 6) The peptide sequence was selected from the N terminal of TSPYL6. Peptide sequence MSLPESPHSPATLDYALEDPHQGQRSREKSKATEVMADMFDGRLEPIVFP.
46 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
Q8N831
DKFZp434B125, testis-specific Y-encoded-like protein 6, TSPY-like 6, TSPY-like protein 6
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
388951
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit