Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565764
Description
Immunogen sequence: TILFLLDPLEG WDSTGDYCLS CLFAQGLPTY TLAVQGISGL PLKKQIDTRK KLSKAVEKRF PHDKLLLLDT QQEAGMLLRQ LANQKQQHLA FRDRRAYLFA HAVDFVPSEE NNLVGTLKIS GYVRGQTLNV NRLLHIVGYG DFQM Highest antigen sequence identity to the following orthologs - mouse 90%, rat 85%.
Required during maturation of the 40S ribosomal subunit in the nucleolus.
Specifications
TSR1 | |
Polyclonal | |
Unconjugated | |
TSR1 | |
AU040765; AW550801; KIAA1401; mKIAA1401; pre-rRNA-processing protein TSR1 homolog; Tsr1; TSR1 20S rRNA accumulation; TSR1, 20S rRNA accumulation, homolog; TSR1, 20S rRNA accumulation, homolog (S. cerevisiae); TSR1, ribosome maturation factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55720 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2NL82 | |
TSR1 | |
Recombinant Human TSR1. Recombinant protein control fragment (Product #RP-104962). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction