Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSSC1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522360

Catalog No. PIPA522360


Only null left
Add to Cart

Description

Description

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (90%), Rat (90%), Zebrafish (83%), Xenopus laevis (82%), Pig (83%), Chicken (92%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TSSC1 has been reported in PMID 9403053 as one of several tumor-suppressing sub transferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alignment of this gene to genomic sequence data suggests that this gene resides on chromosome 2 rather than chromosome 11.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSSC1
Polyclonal
Unconjugated
EIPR1
EARP and GARP complex-interacting protein 1; EIPR1; Endosome-associated recycling protein-interacting protein; Golgi-associated retrograde protein-interacting protein; protein TSSC1; Tssc1; tumor suppressing subtransferable candidate 1; tumor-suppressing STF cDNA 1 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein
Rabbit
Antigen affinity chromatography
RUO
7260
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.75 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q53HC9
EIPR1
Recombinant fragment corresponding to a region within amino acids 1 and 192 of TSSC1 (Uniprot ID#Q53HC9).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.